Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022415.3 | NP_071860.1 | 3 | 1171 | ||
GenBank mRNA | AK010757.1 | - | 3 | 1136 | ||
AK033752.1 | - | 3 | 1136 | |||
AK080897.1 | - | 3 | 1177 | |||
AK150013.1 | - | 3 | 1164 | |||
AK167015.1 | - | 3 | 1040 | |||
BC024960.1 | - | 3 | 1130 | 4950950 |