Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022415.3 | NP_071860.1 | 3 | 2413 | ||
GenBank mRNA | AK080897.1 | - | 3 | 2419 | ||
AK167015.1 | - | 3 | 2282 |