Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCCAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039707.1 | NP_001034796.1 | 4 | 528 | ||
NM_001039708.1 | NP_001034797.1 | 2 | 309 | |||
NM_006643.3 | NP_006634.3 | 3 | 459 | |||
XM_005266050.3 | XP_005266107.1 | 4 | 482 | |||
XM_005266051.3 | XP_005266108.1 | 3 | 373 | |||
XM_011518157.2 | XP_011516459.1 | 3 | 312 | |||
XM_017014218.1 | XP_016869707.1 | 3 | 629 | |||
XM_017014219.1 | XP_016869708.1 | 3 | 629 | |||
GenBank mRNA | AK001296.1 | - | 2 | 88 | ||
AL559847.3 | - | 4 | 462 | |||
AY349357.1 | - | 3 | 149 | |||
BC014515.1 | - | 3 | 326 | 3547130 | ||
BM549484.1 | - | 309 | ||||
BQ723499.1 | - | 316 |