Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022430.2 | NP_071875.2 | 7 | 1044 | ||
GenBank mRNA | AB026044.1 | - | 7 | 994 | ||
AF237906.1 | - | 7 | 982 | |||
AK008099.1 | - | 7 | 983 | |||
AY039038.1 | - | 7 | 982 | |||
BC042719.1 | - | 7 | 1030 |