Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022565.2 | NP_072087.2 | 2 | 1408 | ||
GenBank mRNA | AB036838.1 | - | 1 | 858 | ||
AK140992.1 | - | 2 | 1409 |