Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022722.3 | NP_073559.3 | 10 | 1771 | ||
GenBank mRNA | AK004899.2 | - | 10 | 1768 |