Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271472.1 | NP_001258401.1 | 4, 5 | 627 | ||
NM_022723.3 | NP_073560.2 | 4, 5 | 627 | |||
GenBank mRNA | AF276425.1 | - | 4, 5 | 494 | ||
AK041590.1 | - | 4, 5 | 610 | |||
BC066066.1 | - | 4, 5 | 585 | 6850999 |