Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271472.1 | NP_001258401.1 | 23 | 3940 | ||
NM_001271473.1 | NP_001258402.1 | 20 | 3607 | |||
NM_022723.3 | NP_073560.2 | 22 | 3850 | |||
GenBank mRNA | AF276425.1 | - | 21 | 4159 | ||
AK041590.1 | - | 22 | 3845 | |||
BC066066.1 | - | 23 | 3898 | 6850999 | ||
BC080278.1 | - | 20 | 3575 | 30354974 |