Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199113.1 | NP_001186042.1 | 12 | 1225 | ||
NM_001199114.1 | NP_001186043.1 | 11 | 1064 | |||
NM_001199115.1 | NP_001186044.1 | 11 | 1164 | |||
NM_001199116.1 | NP_001186045.1 | 11 | 1124 | |||
NM_022880.3 | NP_075018.1 | 10 | 1165 | |||
GenBank mRNA | AA407561.1 | - | 5 | 331 | ||
AF131212.1 | - | 10 | 1166 | |||
AF305501.1 | - | 11 | 1135 | |||
AK004756.1 | - | 11 | 1110 | |||
AK050089.1 | - | 11 | 1149 | |||
AK146265.1 | - | 11 | 1065 | |||
AK149483.1 | - | 11 | 1080 | |||
AK170463.1 | - | 11 | 1096 | |||
AK198320.1 | - | 2 | 111 | |||
AK212529.1 | - | 2 | 111 | |||
BC004828.1 | - | 12 | 1188 | 2631728 | ||
BC006812.1 | - | 10 | 1096 | 3155188 |