Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022881.4 | NP_075019.1 | 1 | 289 | ||
GenBank mRNA | AB042807.1 | - | 1 | 304 | ||
AF302685.1 | - | 1 | 295 | |||
AK036407.1 | - | 1 | 290 | |||
BC099387.1 | - | 1 | 284 | 30846527 | ||
BC132544.1 | - | 1 | 290 | 40130821 | ||
BC132546.1 | - | 1 | 290 | 40130823 |