Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164885.1 | NP_001158357.1 | 6, 7 | 1066 | ||
NM_022882.4 | NP_075020.2 | 6, 7 | 942 | |||
GenBank mRNA | AF286723.1 | - | 6, 7 | 942 | ||
AK033662.1 | - | 6, 7 | 979 | |||
AK050018.1 | - | 7, 8 | 1116 | |||
AK136472.1 | - | 1537 | ||||
BC039698.1 | - | 6, 7 | 1055 | 5101211 |