Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164885.1 | NP_001158357.1 | 7 | 1140 | ||
NM_022882.4 | NP_075020.2 | 7 | 1016 | |||
GenBank mRNA | AF286723.1 | - | 7 | 1016 | ||
AK033662.1 | - | 7 | 1053 | |||
AK050018.1 | - | 8 | 1190 | |||
AK136472.1 | - | 1611 | ||||
BC039698.1 | - | 7 | 1129 | 5101211 |