Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199118.1 | NP_001186047.1 | 20 | 3153 | ||
NM_022883.3 | NP_075021.1 | 20 | 3230 | |||
GenBank mRNA | AF286724.1 | - | 20 | 3167 | ||
AK033647.1 | - | 20 | 3181 | |||
AK049363.1 | - | 20 | 3146 | |||
AK163833.1 | - | 20 | 3145 | |||
AK220228.1 | - | 20 | 3230 |