Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022884.2 | NP_075022.2 | 4 | 445 | ||
GenBank mRNA | AF257474.1 | - | 4 | 431 | ||
AK090308.1 | - | 4 | 423 | |||
AK145901.1 | - | 4 | 446 | |||
BC013515.1 | - | 4 | 442 | 4235455 |