Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022886.2 | NP_075024.2 | 3 | 248 | ||
GenBank mRNA | AF245700.2 | - | 3 | 248 | ||
AK020346.1 | - | 3 | 170 | |||
BC111874.1 | - | 3 | 168 | 40059586 | ||
BC112380.1 | - | 3 | 169 | 40059585 | ||
BY775767.1 | - | 3 | 249 |