Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193659.1 | NP_001180588.1 | 4 | 1583 | ||
NM_001193660.1 | NP_001180589.1 | 5 | 1531 | |||
NM_001193661.1 | NP_001180590.1 | 4 | 1418 | |||
NM_022890.2 | NP_075028.1 | 4 | 1573 | |||
GenBank mRNA | AK039672.1 | - | 4 | 1465 | ||
AK049331.1 | - | 4 | 1440 | |||
AK128999.1 | - | 4 | 1452 | |||
AK132155.1 | - | 4 | 1442 | |||
AK166110.1 | - | 4 | 1394 | |||
BC024057.1 | - | 4 | 1482 | 5100104 |