Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022982.2 | NP_075358.1 | 2 | 384 | ||
GenBank mRNA | AF283462.1 | - | 2 | 386 | ||
BC052317.1 | - | 2 | 382 | 6397765 | ||
BC058381.1 | - | 2 | 441 | 6829861 |