Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022999.1 | NP_075375.1 | 7 | 1098 | ||
GenBank mRNA | AK005453.1 | - | 8 | 1142 | ||
AK011086.2 | - | 1 | 365 | |||
BC025098.1 | - | 7 | 1098 | 4159863 |