Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023045.2 | NP_075532.1 | 28 | 4014 | ||
GenBank mRNA | AJ297360.1 | - | 28 | 3919 | ||
AK036837.1 | - | 26 | 3759 | |||
AK039776.1 | - | 1 | 368 | |||
AK122361.1 | - | 27 | 3967 | |||
AK143729.1 | - | 28 | 3985 |