Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clstn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290989.1 | NP_001277918.1 | 6 | 1001 | ||
NM_023051.5 | NP_075538.1 | 7 | 1031 | |||
GenBank mRNA | AJ289016.1 | - | 7 | 862 | ||
AK173059.1 | - | 6 | 1041 | |||
BC029027.1 | - | 2 | 116 | |||
BC053843.1 | - | 6 | 970 | 6315355 | ||
CB525309.1 | - | 4 | 478 |