Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023053.3 | NP_075540.1 | 2 | 261 | ||
GenBank mRNA | AF292033.1 | - | 200 | |||
AF295097.1 | - | 1 | 119 | |||
AJ297390.1 | - | 2 | 211 | |||
AK033504.1 | - | 2 | 220 | |||
AK049646.1 | - | 2 | 255 | |||
AK168547.1 | - | 2 | 211 | |||
AK168633.1 | - | 2 | 220 | |||
BC004850.1 | - | 2 | 216 | 2810960 | ||
CJ165542.1 | - | 2 | 262 |