Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023053.3 | NP_075540.1 | 5 | 3536 | ||
GenBank mRNA | AJ297390.1 | - | 5 | 3484 | ||
AK007481.1 | - | 1 | 495 | |||
AK033504.1 | - | 5 | 3499 | |||
AK049646.1 | - | 5 | 3530 | |||
AK168547.1 | - | 5 | 3486 | |||
AK168633.1 | - | 5 | 3495 | |||
AK190433.1 | - | 1 | 95 | |||
AK205101.1 | - | 1 | 95 |