Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eefsec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023060.3 | NP_075547.1 | 5 | 1022 | ||
GenBank mRNA | AF268871.1 | - | 5 | 986 | ||
AF283518.1 | - | 5 | 977 | |||
AK028424.1 | - | 5 | 1004 | |||
AK087501.1 | - | 5 | 1010 | |||
AK147924.1 | - | 5 | 991 | |||
AK171885.1 | - | 5 | 1002 | |||
AK186069.1 | - | 1 | 217 | |||
BC024915.1 | - | 5 | 996 | 3158026 | ||
BC099675.1 | - | 5 | 980 | 5011896 |