Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lima1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113545.1 | NP_001107017.1 | 6 | 915 | ||
NM_023063.4 | NP_075550.2 | 3 | 787 | |||
GenBank mRNA | AF307844.1 | - | 3 | 710 | ||
AF307845.1 | - | 6 | 869 | |||
AK028186.1 | - | 3 | 728 | |||
AK031698.1 | - | 3 | 731 | |||
AK049350.1 | - | 6 | 865 | |||
AK085065.1 | - | 3 | 781 | |||
AK132314.1 | - | 3 | 733 | |||
AK167220.1 | - | 3 | 347 | |||
BC031490.1 | - | 6 | 841 | 3498138 |