Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023065.3 | NP_075552.2 | 7 | 772 | ||
GenBank mRNA | AK156914.1 | - | 7 | 773 | ||
BC054852.1 | - | 7 | 758 | 6528891 |