Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ethe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023154.3 | NP_075643.1 | 4 | 899 | ||
GenBank mRNA | AB049623.1 | - | 3 | 352 | ||
AK002666.1 | - | 3 | 354 | |||
BC010592.1 | - | 3 | 357 | 3156338 | ||
BC083162.1 | - | 3 | 333 | 6529658 | ||
BC094044.1 | - | 4 | 899 | 5064931 |