Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023167.2 | NP_075656.2 | 9 | 1040 | ||
GenBank mRNA | AB041599.1 | - | 8 | 792 | ||
AB049634.1 | - | 9 | 1039 | |||
AK002464.1 | - | 9 | 876 | |||
AK003746.2 | - | 9 | 997 | |||
BC023078.1 | - | 5 | 2076 | |||
BC037064.1 | - | 9 | 873 | |||
BC039983.1 | - | 9 | 1008 | 4952757 | ||
BC051685.1 | - | 9 | 853 | |||
BC061095.1 | - | 9 | 856 | 6588836 |