Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023178.2 | NP_075667.1 | 3 | 338 | ||
GenBank mRNA | AB221661.1 | - | 3 | 281 | ||
AF265229.1 | - | 3 | 281 | |||
AF438610.1 | - | 3 | 338 | |||
AK005270.1 | - | 4 | 378 | |||
AK012055.1 | - | 4 | 359 | |||
AK136976.1 | - | 2 | 571 | |||
BC002321.1 | - | 3 | 231 | |||
BC023257.1 | - | 3 | 327 | |||
BC045160.1 | - | 3 | 318 | 6309705 |