Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023184.3 | NP_075673.1 | 3 | 2070 | ||
GenBank mRNA | AK007479.1 | - | 2 | 1307 | ||
AK009739.1 | - | 3 | 2069 | |||
AK138824.1 | - | 3 | 2109 | |||
AK142618.1 | - | 3 | 2073 | |||
AK185404.1 | - | 1 | 252 | |||
AK211444.1 | - | 1 | 216 | |||
BC013486.1 | - | 2 | 1990 | 4211067 |