Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023203.1 | NP_075692.1 | 3 | 569 | ||
GenBank mRNA | AF110764.1 | - | 3 | 569 | ||
AK003643.1 | - | 3 | 548 | |||
AK010508.1 | - | 3 | 563 | |||
AK010606.1 | - | 3 | 536 | |||
BC004623.1 | - | 3 | 596 | 3585680 |