Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anp32e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253757.1 | NP_001240686.1 | 7 | 1416 | ||
NM_001253758.1 | NP_001240687.1 | 5 | 1083 | |||
NM_023210.4 | NP_075699.3 | 7 | 1452 | |||
GenBank mRNA | AK049647.1 | - | 7 | 1419 | ||
AK147888.1 | - | 7 | 1418 | |||
AK159446.1 | - | 7 | 1419 | |||
AK168380.1 | - | 7 | 1401 | |||
AK172146.1 | - | 7 | 1466 |