Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023217.4 | NP_075706.1 | 5 | 2079 | ||
GenBank mRNA | AK047835.1 | - | 5 | 2023 | ||
AK049632.1 | - | 5 | 2002 | |||
BC043041.1 | - | 5 | 1989 | |||
BC051938.1 | - | 5 | 2008 | 5717365 |