Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164220.1 | NP_001157692.1 | 3 | 764 | ||
NM_023233.3 | NP_075722.1 | 3 | 702 | |||
GenBank mRNA | AK013959.2 | - | 3 | 674 | ||
AK017850.1 | - | 2 | 659 | |||
AK162129.1 | - | 3 | 752 | |||
BC138576.1 | - | 2 | 628 | 8861596 | ||
BC145915.1 | - | 2 | 628 | 40131218 |