Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168320.1 | NP_001161792.1 | 3 | 239 | ||
NM_006675.4 | NP_006666.1 | 4 | 306 | |||
XM_011520912.2 | XP_011519214.1 | 5 | 423 | |||
XM_011520913.2 | XP_011519215.1 | 2 | 165 | |||
GenBank mRNA | AF089749.1 | - | 4 | 249 | ||
AK027366.1 | - | 4 | 282 | |||
AK223179.1 | - | 4 | 278 | |||
BC071881.1 | - | 3 | 221 | 6256950 | ||
CR542109.1 | - | 2 | 123 |