Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023247.1 | NP_075736.1 | 3, 4 | 761 | ||
GenBank mRNA | AF235017.1 | - | 3, 4 | 761 | ||
AK005252.1 | - | 3, 4 | 376 | |||
AK014634.1 | - | 3, 4 | 376 | |||
BC027513.1 | - | 3, 4 | 794 | 5118121 |