Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023256.1 | NP_075745.1 | 1 | 313 | ||
GenBank mRNA | AF473907.1 | - | 1 | 290 | ||
AK018567.1 | - | 1 | 313 | |||
BC057172.1 | - | 1 | 296 | 4989154 |