Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254761.1 | NP_001241690.1 | 7 | 1903 | ||
NM_023270.5 | NP_075759.3 | 7 | 2029 | |||
GenBank mRNA | AK004847.1 | - | 7 | 2030 | ||
AK146266.1 | - | 7 | 2028 | |||
AK167031.1 | - | 7 | 2016 |