Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nuf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023284.3 | NP_075773.2 | 10 | 916 | ||
GenBank mRNA | AF326732.1 | - | 11 | 957 | ||
AK010351.1 | - | 10 | 838 | |||
AK012011.1 | - | 10 | 850 | |||
AK089939.1 | - | 11 | 944 | |||
AK132467.1 | - | 10 | 917 | |||
AK162249.1 | - | 11 | 952 | |||
BC020026.1 | - | 11 | 925 | 3587655 |