Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290454.1 | NP_001277383.1 | 4 | 674 | ||
NM_023305.3 | NP_075794.3 | 4 | 674 | |||
GenBank mRNA | AF275549.3 | - | 4 | 633 | ||
AK009596.1 | - | 4 | 633 | |||
AK028477.1 | - | 4 | 633 | |||
AK028670.1 | - | 4 | 625 | |||
AK030370.1 | - | 4 | 615 | |||
AK033121.1 | - | 4 | 626 | |||
AK076078.1 | - | 4 | 633 | |||
AK163229.1 | - | 4 | 632 | |||
BC065996.1 | - | 4 | 622 | 5689710 |