Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014811.1 | NP_001014811.1 | 1 | 426 | ||
NM_001161586.1 | NP_001155058.1 | 2 | 499 | |||
NM_006680.2 | NP_006671.2 | 2 | 274 | |||
XM_005273717.1 | XP_005273774.1 | 2 | 375 | |||
XM_017017136.1 | XP_016872625.1 | 2 | 1257 | |||
XM_017017138.1 | XP_016872627.1 | 2 | 282 | |||
GenBank mRNA | AK092181.1 | - | 2 | 237 | ||
AK300974.1 | - | 2 | 499 | |||
AK313695.1 | - | 2 | 349 | |||
BC022472.2 | - | 1 | 426 | 4812045 | ||
BM906572.1 | - | 289 | ||||
DC388623.1 | - | 2 | 499 | |||
X79440.1 | - | 1 | 204 |