Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024177.3 | NP_077139.2 | 4 | 648 | ||
GenBank mRNA | AK088343.1 | - | 4 | 587 | ||
AK151598.1 | - | 4 | 587 | |||
AK159033.1 | - | 4 | 649 | |||
BC002319.1 | - | 3 | 624 | 3593984 |