Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024181.2 | NP_077143.2 | 3 | 836 | ||
GenBank mRNA | AF255459.1 | - | 3 | 513 | ||
AF314002.1 | - | 2 | 349 | |||
AK004617.1 | - | 3 | 591 | |||
AK013930.1 | - | 3 | 532 | |||
AK016269.1 | - | 3 | 836 | |||
AK028184.1 | - | 3 | 576 | |||
AK128945.1 | - | 3 | 591 | |||
AK165652.1 | - | 3 | 594 | |||
AK166502.1 | - | 3 | 546 | |||
AK169020.1 | - | 3 | 603 | |||
BC033461.1 | - | 3 | 575 | 4010341 |