Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Riok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024182.4 | NP_077144.2 | 10 | 1292 | ||
GenBank mRNA | AK004748.1 | - | 10 | 1384 | ||
AK011110.1 | - | 3 | 376 | |||
AK131635.1 | - | 10 | 1296 | |||
AK134327.1 | - | 10 | 1384 | |||
AK147074.1 | - | 10 | 1293 | |||
AK167309.1 | - | 10 | 1388 | |||
AK170882.1 | - | 10 | 1372 | |||
BC002255.1 | - | 7 | 822 | |||
BC033271.1 | - | 10 | 1330 | 4016197 |