Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024188.6 | NP_077150.1 | 4 | 680 | ||
GenBank mRNA | AB085609.1 | - | 4 | 422 | ||
AK009027.1 | - | 4 | 460 | |||
AK010790.1 | - | 4 | 423 | |||
AK011354.1 | - | 4 | 360 | |||
AK078086.1 | - | 4 | 680 | |||
AK146158.1 | - | 4 | 438 | |||
AK146342.1 | - | 4 | 425 | |||
AK151662.1 | - | 4 | 419 | |||
AK167681.1 | - | 4 | 510 | |||
AK178525.1 | - | 4 | 276 | |||
BC003422.1 | - | 4 | 384 | 2598824 | ||
EF095212.1 | - | 1 | 73 |