Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024188.6 | NP_077150.1 | 6 | 921 | ||
GenBank mRNA | AB085609.1 | - | 6 | 663 | ||
AK009027.1 | - | 6 | 701 | |||
AK010790.1 | - | 6 | 664 | |||
AK011354.1 | - | 6 | 601 | |||
AK078086.1 | - | 6 | 921 | |||
AK146158.1 | - | 6 | 679 | |||
AK146342.1 | - | 6 | 666 | |||
AK151662.1 | - | 6 | 660 | |||
AK151695.1 | - | 6 | 651 | |||
AK167681.1 | - | 6 | 751 | |||
AK181314.1 | - | 2 | 136 | |||
AK184597.1 | - | 2 | 137 | |||
AK185295.1 | - | 2 | 137 | |||
BC003422.1 | - | 6 | 625 | 2598824 | ||
EF095212.1 | - | 3 | 314 |