Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024189.6 | NP_077151.3 | 4 | 686 | ||
GenBank mRNA | AB100452.1 | - | 4 | 676 | ||
AB100453.1 | - | 6 | 745 | |||
AK011702.1 | - | 5 | 757 | |||
AK012787.1 | - | 4 | 592 | |||
AK148634.1 | - | 4 | 630 | |||
AK218495.1 | - | 1 | 32 | |||
BC002192.1 | - | 4 | 559 | 3488691 |