Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025321.3 | NP_079597.2 | 6 | 691 | ||
GenBank mRNA | AK002459.1 | - | 6 | 693 | ||
AK012818.1 | - | 6 | 692 | |||
AK032458.1 | - | 693 | ||||
AK135503.1 | - | 6 | 693 | |||
BC005779.1 | - | 6 | 678 | 2810852 | ||
BC083091.1 | - | 6 | 684 | 30445339 |