Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025324.2 | NP_079600.1 | 2 | 504 | ||
GenBank mRNA | AK002594.1 | - | 2 | 500 | ||
AK009053.1 | - | 2 | 501 | |||
AK011620.1 | - | 2 | 505 | |||
AK154328.1 | - | 2 | 511 | |||
BC019995.1 | - | 2 | 517 | 4458724 |