Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025334.3 | NP_079610.1 | 7 | 1182 | ||
GenBank mRNA | AK002862.1 | - | 7 | 1169 | ||
AK003481.1 | - | 7 | 1163 | |||
AK200396.1 | - | 1 | 146 | |||
BC006857.1 | - | 7 | 1182 | 2615934 |