Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167820.1 | NP_001161292.1 | 3 | 767 | ||
NM_001167821.1 | NP_001161293.1 | 2 | 597 | |||
NM_001167822.2 | NP_001161294.1 | 2 | 654 | |||
NM_001167823.1 | NP_001161295.1 | 2 | 670 | |||
NM_001317074.1 | NP_001304003.1 | 2 | 647 | |||
NM_001317075.1 | NP_001304004.1 | 2 | 563 | |||
NM_006698.3 | NP_006689.1 | 2 | 634 | |||
XM_017027599.1 | XP_016883088.1 | 2 | 1547 | |||
GenBank mRNA | AF053470.1 | - | 2 | 573 | ||
AK124822.1 | - | 3 | 492 | |||
AL049288.1 | - | 2 | 580 | |||
BC047692.1 | - | 2 | 565 | 6059195 | ||
BC071704.1 | - | 2 | 559 | 5190373 |